QualaknitESD® KAS Nylon Conductive Knit ESD Gloves, QRP
Supplier: PIP
KASS KASM KASL
76246-442CS 596.92 USD
76246-442 76246-446 76246-448
QualaknitESD® KAS Nylon Conductive Knit ESD Gloves, QRP
Gloves
The QRP KAS Qualaknit ESD Assembly-Inspection Gloves are uncoated gloves for maximum breathability and lowest resistance.
  • Nylon + conductive carbon/nylon yarn
  • Mico-knit fingertipsfor maximum dexterity
  • Uncoated glove for maximum breathability, lowest resistance
KAS Qualaknit ESD Assembly-Inspection Glove is knit from continuous stretch nylon and carbon filiment fibers. Microknit™ fingertip for worker comfort and dexterity. Surface resistivity 108ohms. Seamless knit for greater comfort.