QualakoteESD® PDWS Wave Solder Gloves, ESD-Safe, QRP
Supplier: PIP
PDWSS PDWSM PDWSL PDWSXL
76242-932CS 1705.25 USD
76242-932 76242-934 76242-936 76242-938
QualakoteESD® PDWS Wave Solder Gloves, ESD-Safe, QRP
Gloves
The QRP PDWS Qualakote ESD Low Heat Wave Solder gloves are extra thick for thermal protection.
  • Nylon + conductive carbon/nylon yarn
  • Nitrile foamed palm dip for extra toughness
  • Seamless knit, open back
  • Machine washable for economy of repeated use
PDWS Qualakote ESD Wave Solder Glove is knit from nylon carbon yarn and features a Nitrile anti-static dip palm for dependable grip. Heat protection, yet thin for maximum dexterity. Seamless knit for greater comfort. Surface resistivity 106 ohms.