Pre Pleat™ MERV 8 Pleated Air Filters, Flanders Filters, American Air Filter Company
Supplier: AMERICAN AIR FILTER COMPANY TE
80055.01163 80055.01202 80055.02162 80055.02202 80055.02182 80055.02182 80055.02242 80055.01203 80055.02163 80255.01202 80255.02122 80255.01162 80255.01203 80255.02202 80055.01102 80055.01101 80055.02253 80055.04203 80255.01243 80255.01162 80255.01162 80055.02203 80055.0424A25A 80255.04203
76239-540CS 276.13 USD
76239-540 76239-542 76239-544 76239-546 76239-574 76239-576 76239-578 76239-580 76239-584 76239-590 76239-592 76239-596 76239-622 76239-646 76239-694 76239-884 76239-886 76240-214 76240-370 76240-378 76240-464 76242-608 76245-012 76267-978
Pre Pleat™ MERV 8 Pleated Air Filters, Flanders Filters, American Air Filter Company
Filters Venting Filters
Ecologically advanced filtration medium made entirely from recycled materials.
  • Media maximizing V-pleat design
  • Expanded metal grid prevents media flutter while in operation
  • Diagonal and horizontal support members provide frame strength
  • Filter media pack is sealed to eliminate air bypass
  • Moisture-resistant clay coated frame
The new MERV 8 Pre Pleat 40 filter features a filtering medium that is more efficient and ecologically friendly. Made entirely from recycled materials, this medium achieves MERV 8 efficiency with low resistance to airflow. It isMore Product Information