Powder Peel Red Discs, PR-SH Series, Caplugs
Supplier: Caplugs
PR00312 PR00375 PR00437
76247-220EA 110.82 USD
76247-220 76247-222 76247-224
Powder Peel Red Discs, PR-SH Series, Caplugs
Tapes
The PR-SH Series are die-cut discs made out of our PC11 high temperature polyester film tape used to mask off areas that do not require coating.
  • Recommended for Powder coating, e-coating and anodizing
  • Temperatures up to 425 °F (220 °C)
  • 2 mil polyester, 1.5 mil silicone adhesive
Silicone adhesive will not leave an adhesive residue. PC11 has a higher temperature range than other Polyester tapes.