Polyamide Film Tape, 3M
Supplier: 3M
7000148440 7100007737 7100010443 7010372869 7000050110 7000144792 7000050064 7000050136 7000029143 7000050109 7000036206 5413-3X36YD 7000050109
76238-088CS 3920.71 USD
76238-088 76240-668 76242-352 76242-358 76242-360 76242-362 76242-392 76242-394 76242-396 76243-640 76244-344 76245-606 76245-620
Polyamide Film Tape, 3M
Tapes
3M™ polyimide film tapes does not soften at elevated temperatures, thus the film provides an excellent release surface at elevated temperatures.
  • Dimensionally stable at high temperatures, which helps reduce rework, helping to enable high productivity
  • Flame retardant, chemical and radiation resistant which helps protect surfaces to help reduce replacement costs
  • Polyethylene tape core instead of cardboard
  • Gold tab protection during wave solder of printed circuit boards
3M™ polyimide film tape 5413 with DuPont™ Kapton® polyimide film and siliconeMore Product Information