Saturix Cleanroom Spiral Notebook, College Ruled
Supplier: FG Cleanwipes
Saturix
9716658C CRN-CR-58 971668AC 9716658C
76238-394CS 518.03 USD
76238-394 76244-878 76238-392 76238-544
Saturix Cleanroom Spiral Notebook, College Ruled
Notebooks
Cleanroom notebooks by FG clean wipes are specially designed for use in cleanrooms, these notebooks feature pages impregnated and coated with a unique polymer formulation which inhibits particulate generation and chemical extractables. Control contamination, make cleaning easier, and increase product yield with cleanroom compatible cleanroom notebooks.
  • Compatible with virtually any wet or dry ink
  • Ultra-low particle generation and metallic ion content
  • High brightness and opacity
  • High tear and tensile strength
  • Heat-resistant
  • Clean bagged in an ISO 4 cleanroom environment
Cleanroom notebooks feature college ruled sheets that are spiral-bound and protected with a durable plastic cover. The special anti-particulate polymer formulation applied to each page results in ultra-low particle generation and metallic ion content. Notebooks are recommended for use in ISO 3 to 7 critical environments.