Anti-static ESD Gloves, Renco
Supplier: RENCO CORPORATION TE
RMSG-M R13-RMPG43X R13-RMPG41M R13-RMPG42L R13-RMPG40S
76241-286PK 110.52 USD
76241-286 76243-692 76243-694 76243-696 76243-698
Anti-static ESD Gloves, Renco
Gloves
Anti-static and ESD gloves are widely used in low humidity, flammable and particle sensitive environments. These gloves help protect product and workers from potential dangers.
  • Sturdy and long lasting
  • Cost-effective
  • Powderless
Anti-static gloves are also referred to as Electro-static Dissipative (ESD) gloves, and are ideal for use in electronics cleanrooms, and statically charged environments. These gloves are used while handling electronic applications because static charges can destroy sensitive electronics parts.