Microflex® NeoTouch® 25-201 Neoprene gloves, Ansell
Supplier: Ansell Healthcare
385683 385679 385680 385681 385682
76239-674BX 50.9 USD
76239-674 76239-676 76239-678 76239-680 76239-682
Microflex® NeoTouch® 25-201 Neoprene gloves, Ansell
Gloves
This glove is made with neoprene, an alternative to natural rubber latex that protects against Type I allergies.
  • Powder-free
  • Excellent chemical resistance
  • Superior comfort
  • Roll-beaded cuffs
These gloves provide excellent resistance to acids, bases and alcohols.
Ideal applications include assembly and inspection, lab analysis and technical work, and pharmaceutical manufacturing - especially when resistance to acids is required.
Their exclusive formulation provides superior comfort.
Certifications: This is an exam- and food-grade glove andMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable/Reusable Disposable
Latex/Latex-free Latex-Free
Finish Textured fingers
Cuff Style Extended
Anti-Static +