Green-Lite® Rectangular Combination Ultraviolet/Visible LED Magnifiers, O.C. White
Supplier: O.C. White
Green-Lite®
82400-4-UV-B 82300-4-UV-B
76241-914EA 2223.88 USD
76241-914 76245-070
Green-Lite® Rectangular Combination Ultraviolet/Visible LED Magnifiers, O.C. White
Magnifiers
Equipped with next generation, surface mount LEDs, innovative design create the new standard for illuminated magnifiers.
  • Allows for Ultraviolet Only, Visible only, or combination blend
  • Excellent tool for conformal coating inspection, gem identification, dye penetrant crack inspection
  • Highest lumen output in a lighted magnifier
  • Reduced glare, heat and energy consumption
  • Multi-angle LEDs for shadowless lighting at a full range of focal lengths and heights
  • User controlled LED groups shine desired level of light exactly where required
  • Superb dimming software via convenient, reliable membrane switch
Precision ground, optical grade glass, largest lenses, standard diopter lens.
Low voltage power supplies are auto-switchable. Plug in by simply switching the end power cord. Auto shut off at 4 hour intervals to prevent energy waste while users are away.
High quality die cast aluminum shades and all steel arm components for ESD safety. EZ SWIVEL® Die Cast knuckle allows for precision holding power at any angle. "Elite" arms are slim but have the greatest holding power of any arm of this type in the world. Virtually "maintenance zero".
Certifications: Energy Star compliant (each unit uses 8 W maximum).
Delivery information: Comes with 5 years or 50000 hours pro-rated LED warranty.