Metcal UFTC UltraFine Soldering Cartridges, Conical, OK International
Supplier: OK INTERNATIONAL TE
UFTC-7CNB04 UFTC-7CNL04
76242-186EA 123.49 USD
76242-186 76242-188
Metcal UFTC UltraFine Soldering Cartridges, Conical, OK International
Soldering Supplies Soldering Tips
Conical tip cartridge are for use with the MX-5000/5200 platform with the Metcal UltraFine hand-piece. 700 Series for most standard applications.