ESD-Safe Mat Cleaner, Jensen Tools
Supplier: Jenson Tools
8001
76238-096EA 40.69 USD
76238-096
ESD-Safe Mat Cleaner, Jensen Tools
Cleaning Agents Detergents and Surface Cleansers
Extra strength cleaner for removing dirt, grime and stains from all types of static control surfaces, including table and floor mats, hard laminates, bench tops and portable field service kits.
  • Surface mark remover
  • Fresh, clean scent
  • Water-based liquid
  • Leaves no tacky or corrosive residue