Masking Tape, 3M
Supplier: 3M
7100019551 7000048415 7000047437 7000048382 7010334128 7000124879 7010372825 7000047436 7000088503 7100025511 7000048397 7000127098
76242-378CS 7648.1 USD
76242-378 76242-384 76242-386 76242-458 76243-018 76243-088 76243-634 76244-812 76244-962 76244-966 76245-094 76247-704
Masking Tape, 3M
Tapes
Exceptional non-staining performance on copper, silver and other metals.
  • High temperature resistance for paint-bake applications
  • Synthetic rubber adhesive has moderate adhesion and high heat resistance
  • Crepe paper backing provides easy hand tearing