Thermal Transfer Paper Labels, Z-Select 4000T, Zebra
Supplier: Zebra
Z-Select
LD21TT1P5
76253-030CS 545.1 USD
76253-030
Thermal Transfer Paper Labels, Z-Select 4000T, Zebra
Labels
The Zebra Z-Select 4000T is a thermal transfer, bright white, coated, ultra-smooth, paper label with a permanent acrylic adhesive. Specially coated for use with compatible Zebra ribbons for superior durability and image quality.
  • Offers excellent adhesion on corrugated packaging
Recommended for a wide variety of applications, such as shipping, receiving, and work-in-process labels. These labels offer excellent print quality for a variety of general-purpose applications such as general office, barcode, shipping, asset tracking, bakery, weigh scale, industrial, retail, produce, shelf marking, pharmaceutical, and point of sale.
Packaging: 2500 labels per roll; 8 rolls per case.