2-Propanol 70% (v/v)

Supplier: ANDWIN SCIENTIFIC

Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-Propanolsec-Propyl Alcohol2-Hydroxy propaneDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

LC15760-4 LC15760-5 LC15760-200
76238-872EA 175.01 USD
76238-872 76243-476 76238-874
2-Propanol 70% (v/v)
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
MDL Number: MFCD00011674
CAS Number: 67-63-0
https://us.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR