Inocculating Loops, McKesson Medical-Surgical
Supplier: MCKESSON MEDICAL-SURGICAL INC TE
16-1RL COP-H10
76238-538PK 248.25 USD
76238-538 76238-540
Inocculating Loops, McKesson Medical-Surgical
Spreaders Inoculating Loops and Needles
Inocculating Loops are color-coded for easy product identification and hexagonal loop shaft for easy handling.
  • Ultra smooth surface for perfect streaking
  • Free of lubricants, oils and electrostatic charges
  • Loop accuracy certified using Evans blue dye method
  • Tamperproof ziplock resealable bag
Certifications: Certification of calibration included.
Order Now
SPECIFICATIONS
Environmentally Preferable