Anti-Static Poly Bags, Uline
Supplier: Uline
S-12557
76244-478PK 363.98 USD
76244-478
Anti-Static Poly Bags, Uline
Bags Storage Bags
These poly bags are anti-static and non corrosive.
  • Made of virgin polyethylene film
  • Heat sealable with impulse sealers