Latex Rolled Fingercot, Calapro
Supplier: Calapro
FY01-XL
76238-772CS 1729.14 USD
76238-772
Latex Rolled Fingercot, Calapro
Finger Cots Latex Finger Cots
Latex rolled fingercot processed by advanced chlorination washing to minimize contamination both inside and outside.
  • Powder free
  • Inspected and packaged under class 100 specifications