Essentra Wipes C30, FG Clean Wipes
Supplier: FG Cleanwipes
7C3022P00 7C3044P00 C30-99 7C30PPP00 7-C30B-99P-00
76243-588CS 1161.67 USD
76243-588 76238-520 76243-606 76238-522 76238-336
Essentra Wipes C30, FG Clean Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
FG clean wipes are pioneer in the manufacturing of wipes for cleanrooms.
  • Economical
  • Low NVRs and ions
  • Super absorbent
  • Extremely low particle generation
  • No chemical binders
Wipes are hydroentangled and made of 55% cellulose and 45% polyester.
Suitable for ISO 5, 6, 7 and 8 environments.