Medical / Laboratory ESD-Safe Fabric Chair, Gibo Kodama
Supplier: GIBO KODAMA TE
EIN51IR-F851-07
76243-104EA 3803.2 USD
76243-104
Medical / Laboratory ESD-Safe Fabric Chair, Gibo Kodama
Chairs
Adjustable ESD safe fabric chair with aluminum base and saddle seat.
  • Autonomous tilt control with seat and back angle adjustments
  • Chrome plated cylinder column
  • Saddle seat
Surface resistance: 106 Ω/sq
Seat adjustable from 17 - 211/2"