Engraver Kit, Dremel
Supplier: Dremel
290
76252-190EA 132.03 USD
76252-190
Engraver Kit, Dremel
Power Tools
This unique kit suitable for engrave or decorate a wide variety of materials, including metal, plastic, glass, ceramic, wood and leather.
  • Stroke adjustment dial regulates engraving depth from fine lines to deep grooves
  • Replaceable carbide point lets you engrave on most surfaces
  • Optional 9929 diamond point (not included) for heavy-duty use on hard surfaces
  • Letter/number template helps etch words and numbers
  • Soft grip adds extra comfort and control
  • Light and compact design provides comfortable use for precision engraving performance
Delivery information: Kit includes a tool with a soft-grip body and variable stroke control so you can etch both fine lines and deep grooves. Also included is a replaceable carbide engraving point.