Pliers Set, Wiha Tools
Supplier: WIHA TOOLS TE
32685
76248-482EA 110.5 USD
76248-482
Pliers Set, Wiha Tools
Pliers
These pliers are D shaped and have 90° angled tips.
  • Die forged cylindrical precision formed
  • Made Chromium-vanadium special tool steel
  • Soft vinyl grips
Ideal for inserting and removing internal retaining rings.
Delivery information: Set includes pliers have sizes as follows, 8 to 13 mm with 0.035" tip, 12 to 25 mm with 0.050" tip, 19 to 60 mm with 0.070" tip, 40 to 100 mm with 0.090" tip and 85 to 140 mm with 0.126" tip.