Fisnar® Dispensing Needles, Ellsworth Adhesives
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
5601130
76245-062PK 94.89 USD
76245-062
Fisnar® Dispensing Needles, Ellsworth Adhesives
Syringe Needles
These dispensing needles are straight in shape and tips are blunt end.
  • Polished burr-free finish for smooth fluid dispense
Made of stainless steel and epoxy sealed to a double helix Luer lock polypropylene hub.