Thin Nose Diagonal Cutter, Jensen Tools + Supply
Supplier: Jensen Tools + Supply
95135
76248-628EA 22.93 USD
76248-628
Thin Nose Diagonal Cutter, Jensen Tools + Supply
Cutters
Thin nosed cutter features precision flush cutting jaws with thin tapered nose for working in tight spaces.
  • Handles are spring loaded
  • Not for use on live circuits
Certifications: Meet ANSI specifications.