Chlorine Free Medium Duty Body Bags, Extra Packaging Corporation
Supplier: EXTRA PACKAGING CORP. TE
BBCS-50-CF
76238-472CS 427.35 USD
76238-472
Chlorine Free Medium Duty Body Bags, Extra Packaging Corporation
Bags Emergency Bags
These bags feature two layers of 3-mil polyethylene film laminated to 1000 denier scrim reinforced material.
  • Electronic heat-sealed construction
  • No vinyl material used in manufacturing
  • Adhesive tape backed reinforced zipper to minimize possible zipper tear-out during handling and transport
Chlorine free material remains flexible to −70 °F, so that it can be used in winter environments or morgue coolers without fear of material cracking.
Certifications: Comply with OSHA regulation 3130.
Ordering information: Each bag packed with BBID-KIT-T for field identification without the hassle of carrying separate ID tags and supplies. Kit consists of tear proof Tyvek toe tags, exterior ID tag, personal effects ID tag, red biohazard tag and yellow contamination tags.