Chart Recorder, L200E, Linseis
Supplier: Kipp & Zonen
L120/5 LO/8041 L200E-1
76247-108PK 290.7 USD
76247-108 76247-110 76247-574
Chart Recorder, L200E, Linseis
Chart Recorders Recorders
Deluxe Flatbed Chart Recorder built according to CE and FCC regulations. Single channel.
  • Chart width 200 mm
  • mV, V and mA inputs
  • Separate electric pen lift per channel
  • Chart drive with grid function
  • 16 chart speeds from 0,1 mm/min to 20 mm/sec
  • Remote control for chart drive
  • Single chip hybrid technology
  • Long life fibre tip pens
  • Monitor output signal
  • Paper advance/rewind knob