SmartRack 3U Fan Panel, Global Industrial
Supplier: Global
SRFAN3U
76250-016EA 790.26 USD
76250-016
SmartRack 3U Fan Panel, Global Industrial
Ventilators
Tripp Lite's SmartRack Panel-mount Triple Fan Accessory is ideal for increasing the ventilation of SmartRacks, enclosures and server cabinets.
  • The unit features (3) 4", 120 V fans mounted in a 3U panel, 6 feet cord
Delivery information: It includes mounting hardware.