3M™ Attest™ Biological Indicator, NDC
Supplier: NDC
1264-S
76241-072BX 2056.25 USD
76241-072
3M™ Attest™ Biological Indicator, NDC
Sterilization Indicators
Indicator designed for reliable monitoring for ethylene oxide sterilization process when used in conjunction with an appropriate biological indicator incubator and life science/ industrial sterilization applications.
  • 48-hour standard readout
  • Self-contained design
This is comprised of a paper carrier inoculated with a standardized population of Bacillus atrophaeus ATCC® 9372 spores, a glass ampule containing recovery medium which includes a pH indicator dye, a plastic cap with filter, and a plastic vial. The biological indicator has a chemical process indicator on the label that will change from reddish-brown to green upon exposure to the ethylene oxide sterilization process. The recovery media will produce a pH color change from blue-green to yellow if viable spores are present and incubation conditions are correct. Also verified with a readout or Reduced Incubation Time (RIT) of 48 hours under specific test conditions.
Certifications: Compliant with ANSI/AAMI/ISO 11138:(2006) Parts 1 and 2