Approved Power Supplies, 0 to 30 VDC
Supplier: OSAW INDUSTRIAL PRODUCTS SE
SE1094 SE1096 SE1095
470235-886CT 70.2 USD
470235-886 470235-884 470235-882
Approved Power Supplies, 0 to 30 VDC
Power Supplies
A high-quality, regulated power supply for the laboratory.
  • Regulated continuous independent voltage and current adjusting knobs
  • Thermal protection
  • Digital display
  • Lockable knobs
0 to 30 VDC regulated and continously variable power supplies with independent voltage and current knobs.