Vacuum Filtration Adapters, with Pluro Stopper Flared Tops, Chemglass
Supplier: Chemglass
CG-1053-11 CG-1053-13 CG-1053-16
80066-088EA 130.87 USD
80066-088 80066-092 80066-098
Vacuum Filtration Adapters, with Pluro Stopper Flared Tops, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter used for reduced pressure filtration with plain stem buchner funnels.
  • Top is flared to accept the listed size Pluro stopper
  • The hose connection has an O.D. of 10 mm at the largest serration
Delivery information: Adapters are not supplied with stoppers.