Distillation Adapters, Chemglass
Supplier: Chemglass
CG-1027-01 CG-1027-14
80065-858EA 121.85 USD
80065-858 80065-862
Distillation Adapters, Chemglass
Distillation Components Distillation Adapters
Adapters have a #7 Chem-Thread at the top for a vacuum-tight seal of plain stem thermometers.
Adapters feature the same size inner joints on the bottom and sidearm at a 75° angle to one another.
Delivery information: Supplied with a compression cap and Viton O-Ring