Miller® Durahoist™ Portable Confined Space Systems, Honeywell Safety
Supplier: Honeywell Safety
Miller®
DH-1/ DH-2/
89400-612EA 12279.23 USD
89400-612 89400-614
Miller® Durahoist™ Portable Confined Space Systems, Honeywell Safety
Fall Protection Fall Arresters
The Miller DuraHoist™ portable confined space system is designed for manhole and confined space entry/retrieval, and fall arrest applications.
  • System breaks down into four lightweight aluminum components
  • No tools required for assembly
The modular design allows the use of components in various configurations and applications, such as fixed and portable bases and an optional mast with extensions. Multiple winches and/or lifeline systems may be mounted as required. The lightweight, manageable components are easy to store, transport, and set-up.