Modified Claisen Adapters, Chemglass
Supplier: Chemglass
CG-1074-01 CG-1074-02 CG-1074-14
80066-176EA 263.33 USD
80066-176 80066-178 80066-180
Modified Claisen Adapters, Chemglass
Distillation Components Distillation Adapters
Claisen-style adapters with an additional standard taper outer joint at a 45° angle to the vertical outer joint.