Jenner's stain stock solution for H-Pylori

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Eosin−Methylene Blue

26114-01
100505-038EA 38.88 USD
100505-038
Jenner's stain stock solution for H-Pylori
Jenner's stain

Used May-Grunwald Giemsa for H-Pylori

Flash Pt: 11 °C (51.8 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081733
CAS Number: 62851-42-7
EINECS: 200-659-6
UN: 1230
ADR: 3, 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR