DYNEO™ DD Refrigerated/Heating Circulators
Supplier: Julabo
DYNEO™
9021708.33.A 9021701.33 9021714.N1.02.A 9021701.02.D 9021701.33.A 9021701.02 9021703.02.D 9021705.02.A 9021702.33.A 9021714.S1.02 9021705.33.A 9021713.N1.33.A 9021701.N1.02.A 9021701.02 9021703.13 9021702.02 9021707.33.A 9021714.N1.02 9021703.02 9021704.02.A 9021704.02 9021713.N1.02.D 9021704.33 9021706.33 9021701.N1.02.D 9021705.02.A 9021714.S1.33.D 9021714.N1.02.D 9021728.N1.02 9021702.33 9021701.N1.33 9021701.33.D 9021705.33.D 9021701.02.A 9021703.13 9021708.33.D 9021717.N1.33.A 9021717.N1.02.A 9021728.N1.02.A 9021717.N1.33.D 9021713.S1.33.A 9021702.02.D 9021727.33.D 9021704.02 9021707.33 9021707.02.D 9021701.02.A 9021713.N1.02.A 9021701.02.D 9021714.N1.33.D 9021702.33.D 9021707.33.D 9021713.N1.33.D 9021714.N1.33.A 9021706.33 9021707.02.A 9021728.N1.33.D 9021701.N1.33.D 9021713.S1.33.D 9021708.33.D 9021704.33.D 9021713.S1.02.D 9021708.33 9021705.02 9021706.02.A 9021713.S1.33 9021705.33 9021707.33.A 9021727.02.D 9021708.33 9021727.33.A 9021715.N1.02 9021705.02 9021705.33 9021713.S1.02 9021705.33.D 9021704.02.D 9021707.33 9021703.13.D 9021702.02.D 9021703.13.A 9021715.N1.02.D 9021705.33.A 9021706.33.D 9021702.33.A 9021706.02.A 9021707.02 9021717.N1.33 9021702.33.D 9021703.02.A 9021728.N1.33.A 9021708.33.A 9021713.N1.02 9021727.02 9021701.33 9021706.02 9021701.33.A 9021713.N1.33 9021714.S1.02.D 9021704.33.A 9021715.N1.33.D 9021714.S1.02.A 9021702.02.A 9021704.02.D 9021706.02.D 9021702.02 9021728.N1.02.D 9021727.02.A 9021706.02.D 9021706.33.D 9021714.S1.33 9021702.02.A 9021714.N1.33 9021705.02.D 9021715.N1.33 9021706.02 9021703.13.A 9021707.02.A 9021717.N1.02 9021707.33.D 9021714.S1.33.A 9021704.33.D 9021705.02.D 9021704.02.A 9021701.N1.33.A 9021715.N1.33.A 9021707.02.D 9021703.02 9021702.33 9021703.02.D 9021707.02 9021704.33.A 9021706.33.A 9021701.N1.02 9021703.02.A 9021701.33.D 9021713.S1.02.A 9021715.N1.02.A 9021717.N1.02.D 9021706.33.A 9021704.33 9021728.N1.33 9021727.33 9021703.13.D
76289-292EA 9666.19 USD
76289-292 76558-184 76289-290 76558-186 76289-296 76289-330 76289-294 76289-334 CA76289-342 76289-298 CA76289-340 76289-332 76289-338 CA76289-346 76558-182 CA76289-344 76289-336 77442-104 CA76289-338 77442-102 76289-328 77442-108 77442-106 76558-214 77442-100 76558-172 76289-282 CA76289-290 76558-176 76289-286 CA76289-294 76558-178 76289-284 CA76289-292 CA76289-332 76289-322 CA76289-298 CA76289-330 76289-320 76289-288 CA76289-296 CA76289-336 76289-326 76558-170 76289-324 CA76289-334 CA76289-328 CA76289-326 76289-318 76558-202 76558-204 76558-208 76558-162 76558-164 CA76289-284 CA76289-282 CA76289-288 CA76289-320 76289-312 CA76289-286 76289-310 76289-316 CA76289-324 CA76289-322 76289-314 76289-308 CA76289-318 CA76289-316 76289-306 76558-158 76289-380 76558-274 76289-384 76289-382 CA76289-310 76289-300 76289-304 CA76289-314 CA76289-312 76289-302 CA76289-306 CA76289-304 CA76289-308 76558-260 76558-264 CA76289-382 76289-374 76558-266 CA76289-380 76289-378 CA76289-384 76289-376 CA76289-302 CA76289-300 76558-258 76558-250 76558-252 77442-092 76558-256 CA76289-376 CA76289-374 CA76289-378 77442-138 77442-136 77442-096 77442-130 77442-094 76558-248 77442-134 77442-098 77442-132 76558-242 76289-352 76558-244 76289-350 76289-356 76289-354 76289-358 77442-126 77442-124 77442-128 77442-122 77442-120 76558-230 CA76289-350 76289-340 76558-232 76289-344 CA76289-354 CA76289-352 76289-342 CA76289-358 76289-348 CA76289-356 76558-192 76289-346 77442-116 CA76289-348 77442-114 77442-118 76558-224 76558-226 77442-112 77442-110
DYNEO™ DD Refrigerated/Heating Circulators
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
DYNEO™ DD refrigerated/heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from –20 to 200 to –50 to 200 °C.
  • Infinitely adjustable pressure pump
  • Easy switching between internal and external circulation
  • Large color TFT display, multilingual interface
  • Integrated programmer
  • PT100 port facilitates external temperature control via PT100 sensor
  • USB port
  • Optional RS232 interface or analog interfaces
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
DYNEO DD circulator features a color TFT display with central knob controller for operating/programming. Integrated programmer. A lever below the display enables simple adjustment between internal and external fluid flow. The USB port facilitates external communication with models options to add RS232 interface or analog interface. Operation in °C or °F. Pump connections: M16×1 male threads.
Certifications: CE, UKCA. DIN 12876-1 Class III.
Ordering information: Each unit is supplied with a pair of 8 and 12 mm hose barbs. Fluid, adapters, tubing sold separately.
Delivery information: Includes DYNEO DD immersion circulator, refrigeration unit, mounting bridge, bath tank lid, pair of 8 and 12 mm hose barbs, integrated drain tap.