Glass Beads, Biospec Products
Supplier: BIOSPEC PRODUCTS MS
11079127 11079135 11079635 11079105
MSPP-11079127EA 45.23 USD
MSPP-11079127 MSPP-11079135 MSPP-11079635 MSPP-11079105
Glass Beads, Biospec Products
Beads Glass Beads
Glass has a density of 2.5 g/cc and is the most commonly used bead media for beadbeating.
  • Grinding grade
Beads are sold in one pound bottles with easy pour spouts. Beads are sorted by sieves, bead size varies between ±10%.