Soldering Tips for NP105 and NT105 Nano Tools, JBC
Supplier: JBC TOOLS USA INC. TE
C105101 C105105 C105108 C105103 C105112 C105113 C105106 C105110 C105114 C105111 C105107 C105213 C105214 C105118 C105120 C105116 C105126 C105211 C105222 C105223 C105125 C105124 C105117
76240-992EA 175.3 USD
76240-992 76240-994 76240-996 76240-998 76243-170 76243-196 76244-316 76244-852 76245-176 76245-294 76245-556 76245-630 76245-632 76245-692 76245-890 76246-764 76248-928 76250-748 76254-342 76254-344 76268-064 76277-310 76277-022
Soldering Tips for NP105 and NT105 Nano Tools, JBC
Soldering Supplies Soldering Tips
The Galvanic treatment of tips offers outstanding thermal performance and long duration.
The C105 cartridges are appropriate for soldering and repairing very small-sized components at high precision, even under a microscope.
C105 works with NT105 Nano Soldering Iron and NP105 Nano Tweezers. It also features exclusive heating system and the sleep and hibernation options.
Delivery information: Supplied with 5 units in the DC-A dispenser case.
Order Now
SPECIFICATIONS
Environmentally Preferable