Bovine Pituitary Extract, Pel-Freez Biologicals
Supplier: PEL-FREEZ BIOLOGICALS MS
57136-3 57136-2
100372-394EA 2286.15 USD
100372-394 100372-392
Bovine Pituitary Extract, Pel-Freez Biologicals
Cell Culture Supplements and Additives
Fresh frozen pituitaries are obtained from normal healthy bovine cattle.
  • Useful as a supplement to culture a variety of epithelial or endothelial cells
The pituitaries are extracted using a saline extraction and the extract is filtered through a 0.2 micron filter. The extract remains at or below +5 °C throughout the procedure until the product is frozen. The tissue is sourced from a USDA certified facility and the animals receive ante and post mortem inspection and found to be free of contagious diseases.