Silcon® Med-X Platinum Cured Silicone Tubing, Newage Industries
Supplier: NewAge Industries
284-1482-50 284-1286-50 284-1279-50 284-1580-50
76241-198EA 503.59 USD
76241-198 76241-208 76241-234 76241-236
Silcon® Med-X Platinum Cured Silicone Tubing, Newage Industries
Tubing Transfer Tubing
Silicone tubing made from platinum cured for the highest degree of purity.
  • Reusable will withstand repeated sterilization
  • Free of BPA, latex and phthalates
Surface properties resist sticking and encrustation and will not support bacteria growth. The tubing is soft, pliable, and contains no plasticizers which could leach out, causing flow contamination or tube hardening.
Applications include pharmaceutical, biomedical, health and beauty, food, and beverage handling.
Certifications: Conforms to the biocompatibility requirements of USP Class VI.
Packaging: Supplied individual in heat-sealed polybags.
Caution: Not recommended for implantable or in-body uses or for continuous steam applications.