Circlegrow™ Powder, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
113000122 113000132 113000142 113000152
76019-864EA 162.17 USD
76019-864 76019-868 76019-870 76019-872
Circlegrow™ Powder, MP Biomedicals
Cell Culture Media
Obtain rapid growth of E. coli and high yields of plasmid DNA past log phase.
  • Proprietary media for enhanced cell and plasmid yield
  • Convenient packaging formats for a variety of applications and budgets
  • Proven and reliable reputation
  • High-quality raw materials and additives
Circlegrow™ media is designed for obtaining rapid growth of E. coli and high yields of plasmid DNA. Bacteria grown in conventional culture media reach an abrupt stationary phase upon exhaustion of nutrients. Cell proliferation is halted completely, and some cell death and plasmid loss occur. At the end of log phase, cells growing in Circlegrow™ begin to utilize a proprietary second carbon source that becomes available on a delayed basis. Cell growth and plasmid replication continue long after cells in conventional media have reached stationary phase or expired. Bacteria commonly grow in Circlegrow™ to a density triple that achieved in other media. The slow post-log phase growth of bacteria in Circlegrow™ eliminates the need for constant OD monitoring. Circlegrow™ cultures left overnight or over the weekend will have a much lower percentage of dead cells, and a higher percentage of plasmid DNA, than cultures prepared with any other media. The result is a higher yield of plasmid DNA in less volume with less attention.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.
Certifications: Molecular Biology Certified™.
Caution: Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media.