Handles for König® Green Band Fiber Optic Laryngoscopes Blades
Supplier: MEDLINE INDUSTRIES, INC.
MDS0423615 MDS0423625 MDS0423820
470303-280EA 221.8 USD
470303-280 470303-276 470303-278
Handles for König® Green Band Fiber Optic Laryngoscopes Blades
Razors and Blades
These knurled handle provides secure grip for the fiber optic laryngoscopes.
  • LED or xenon bulb provides bright, white illumination
The blade/handle combination provides 2X illumination of other systems.
Ordering information: Xenon bulb replacement available.