Orthophosphoric acid 50% (v/v)

Supplier: Ricca Chemical
Danger

Synonyms: Phosphoric acid

R5853000-20F R5853000-4A
RCR5853000-20FEA 1250 USD
RCR5853000-20F RCR5853000-4A
Orthophosphoric acid 50% (v/v)
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Thymolphthalein) 49.5-50.5 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR