FastGene Agarose Gel Band Cutter, Bulldog Bio
Supplier: Bulldog Bio
FG-830
102407-950EA 90.77 USD
102407-950
FastGene Agarose Gel Band Cutter, Bulldog Bio
Cutters
All-in-One Disposable Exciser and Dispenser.
  • This affordable tool simplifies fragment purification and eliminates wasted effort using Exacto knives or razor blades
  • The FastGene Agarose Gel Band Cutter is a ready-to-use, disposable tool for cutting agarose gel bands
Manually excising gel bands takes time and practice, but now there’s another way to easily cut out DNA bands with both consistency and accuracy.
The size of the excised gel band will always be 6×3 mm, and you can collect multiple bands in a single FastGene Cutter—making large-scale purifications that much easier.