Platinum, disc, drilled apertures 6.35 mm O.D, Thickness 0.14 mm
Supplier: Electron Microscopy Sciences
Synonyms:
Platinium
Platinum apertures are easily cleaned by either an aperture flamer or over a bunsen burner in the open air. We maintain a wide variety of Platinum Drilled Apertures in stock with different configurations suiting all of your needs.
Platinum Apertures size tolerances vary as follow:
• Nominal hole diameter: +/- 1 micron
• Disc diameter: +/- 10 microns
• Disc thickness: +/- 10 microns
Platinum Apertures size tolerances vary as follow:
Hole Sizes not listed are available upon special request.
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...