Copper, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91006-CU 91007-CU 91008-CU 91009-CU 91010-CU 91013-CU 91014-CU 91015-CU 91016-CU 91017-CU 91018-CU 91019-CU 91020-CU
102099-402EA 189.27 USD
102099-402 102099-432 102099-462 102099-492 102099-520 102099-552 102099-582 102099-612 102099-642 102099-672 102099-702 102099-732 102099-762
Copper, sputtering target sputter coater cathodes
Copper

EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.


EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.

Formula: Cu
MDL Number: MFCD00010965
CAS Number: 7440-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR