Silver, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91006-AG 91007-AG 91008-AG 91009-AG 91010-AG 91013-AG 91014-AG 91015-AG 91016-AG 91017-AG 91018-AG 91019-AG 91020-AG
102099-394EA 486.13 USD
102099-394 102099-424 102099-454 102099-484 102099-514 102099-544 102099-574 102099-604 102099-634 102099-664 102099-694 102099-724 102099-754
Silver, sputtering target sputter coater cathodes
Silver

EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.


EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.

Formula: Ag
MDL Number: MFCD00003397
CAS Number: 7440-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR