Silicon carbide, abrasive film discs, Diameter 8" PSA Back

Supplier: Electron Microscopy Sciences

50258-1 50258-15 50258-3 50258-30 50258-5 50258-60 50258-9
102092-768PK 103.75 USD
102092-768 102092-770 102092-772 102092-774 102092-776 102092-778 102092-780
Silicon carbide, abrasive film discs, Diameter 8" PSA Back
Silicon carbide
Formula: SiC
MW: 40.1 g/mol
MDL Number: MFCD00049531
CAS Number: 409-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR