Magid® Economy Size Lens Cleaning Station, Magid Glove
Supplier: Magid Glove
M300D M200D
32893-191EA 39.46 USD
32893-191 32893-201
Magid® Economy Size Lens Cleaning Station, Magid Glove
Wipes Disposable Wipes and Towels Lens Papers
Magid's line of economy size lens cleaning stations come with disposable tissues and lens cleaning pumps.
  • Anti-fog, anit-static liquid lens cleaner and disposable low-linting, non-abrasive tissues
  • Large 5×8", interfolded tissues
  • Use for cleaning glasses, goggles, face shields, respirators
  • Pump dispenser
Perfect for cleaning glass, plastic and polycarbonate lenses and surfaces. Completely assembled, ready-to-use with pre-cut openings for easy hanging.
Ordering information: Stations available in small (8 oz. dispenser) and large (16 oz. dispenser).