Magid® CutMaster® Medium Weight Kevlar® Leather Palm Gloves, Magid
Supplier: Magid Glove
KEV16LEA9 KEV16LEA7
69100-168CS 2741.32 USD
69100-168 69100-600
Magid® CutMaster® Medium Weight Kevlar® Leather Palm Gloves, Magid
Gloves
This gloves combines the toughness and durability of a split leather palm with a 100% Kevlar machine knit shell.
  • Quality cow split leather palm is sewn on using Kevlar thread to ensure a long-lasting seam
  • Color-coded overedge wrist prevents unraveling and keeps a tight fit after washing
Kevlar fiber is five times stronger than steel and gives this glove superior ANSI cut Level 4 protection.