Magid® D-ROC® Hyperon™ Palm Coated Gloves, Magid
Supplier: Magid Glove
PF5406 PF5407 PF5408 PF5409
69100-291PK 20.93 USD
69100-291 69100-293 69100-295 69100-297
Magid® D-ROC® Hyperon™ Palm Coated Gloves, Magid
Gloves
The D-ROC® gloves feature the cut resistance of a Hyperon™ blended machine knit shell with the added abrasion resistance of a polyurethane coated palm. The lightweight, form-fitting design of the multi-purpose work gloves provides excellent comfort while delivering superior grip, dexterity.
  • Blended shell provides excellent comfort and dexterity, and 360° cut protection
  • Polyurethane coating channels away oils and ensures a secure grip
  • Machine washable for longer life
  • Color-coded overcast acts as a visual aid for sizing and reduces fraying
  • Cut and abrasion resistance
Low-linting, the fitted gloves have a shell that is finished with a rubber band cuff to prevent unravelling.
Certifications: ANSI Level 2 cut protection.