Platinum 5% on calcium carbonate powder, dry 3 µm for use as a catalyst, unreduced , BASF Catalyst

Supplier: Spectrum Chemical Mfg. Corp.

SP-205-100G SP-205-25G SP-205-5G
76124-502EA 1727.55 USD
76124-502 76124-504 76124-506
Platinum 5% on calcium carbonate powder, dry 3 µm for use as a catalyst, unreduced , BASF Catalyst
Platinum

5 Percent Platinum on Calcium Carbonate Powder, Unreduced, Dry, 3 microns, BASF Catalyst

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR